missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-2,6-Dimethylpiperidine, 97+%, ACROS Organics™

Manufacturer:  Acros Organics 116681000

 View more versions of this product

Catalog No. AC116681000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

cis-2, 6-Dimethylpiperidine
Authentic
1.4380 to 1.4420 (20°C, 589nm)
100g
766-17-6
100.0
C7H15N
2733
02,156
SDGKUVSVPIIUCF-KNVOCYPGSA-N
(2R,6S)-2,6-dimethylpiperidine
641768
Liquid
0.8400g/mL
Glass bottle
Colorless to Yellow
99%, min. 95% cis-isomer
97.0
97+%
MFCD00005986
20, 108
cis-2,6-dimethylpiperidine, 2r,6s-2,6-dimethylpiperidine, cis-hexahydro-2,6-lutidine, cis-2,6-lupetidine, piperidin, 2e,6e-dimethyl, cis-2,6-dimethyl-piperidine, cis-lupetidine, cis-nanophine, prestwick0_000666, prestwick1_000666
CC1CCCC(N1)C
113.204
113.2
≥97.0%

Chemical Identifiers

766-17-6
113.204
SDGKUVSVPIIUCF-KNVOCYPGSA-N
641768
CC1CCCC(N1)C
C7H15N
MFCD00005986
cis-2,6-dimethylpiperidine, 2r,6s-2,6-dimethylpiperidine, cis-hexahydro-2,6-lutidine, cis-2,6-lupetidine, piperidin, 2e,6e-dimethyl, cis-2,6-dimethyl-piperidine, cis-lupetidine, cis-nanophine, prestwick0_000666, prestwick1_000666
(2R,6S)-2,6-dimethylpiperidine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA

SDS Documents