missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
cis-2,6-Dimethylpiperidine, 97+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 116680050
Specifications
cis-2, 6-Dimethylpiperidine | |
11°C | |
Glass bottle | |
0.84 | |
Colorless to Yellow | |
99%, min. 95% cis-isomer | |
97.0 | |
97% min. (GC) | |
MFCD00005986 | |
20, 108 | |
2r,6s-2,6-dimethylpiperidine, cis-2,6-dimethyl-piperidine, cis-2,6-dimethylpiperidine, cis-2,6-lupetidine, cis-hexahydro-2,6-lutidine, cis-lupetidine, cis-nanophine, piperidin, 2e,6e-dimethyl, prestwick0_000666, prestwick1_000666 | |
SDGKUVSVPIIUCF-KNVOCYPGSA-N | |
(2R,6S)-2,6-dimethylpiperidine | |
641768 | |
Liquid |
0.8400g/mL | |
Authentic | |
1.4384 to 1.4404 | |
127°C to 128°C | |
5g | |
766-17-6 | |
100.0 | |
C7H15N | |
2733 | |
02,156 | |
Solubility in water: miscible. Other solubilities: soluble in most organic solvents | |
CC1CCCC(N1)C | |
113.2 | |
113.2 | |
97+% |
Chemical Identifiers
766-17-6 | |
113.2 | |
SDGKUVSVPIIUCF-KNVOCYPGSA-N | |
641768 | |
CC1CCCC(N1)C |
C7H15N | |
MFCD00005986 | |
2r,6s-2,6-dimethylpiperidine, cis-2,6-dimethyl-piperidine, cis-2,6-dimethylpiperidine, cis-2,6-lupetidine, cis-hexahydro-2,6-lutidine, cis-lupetidine, cis-nanophine, piperidin, 2e,6e-dimethyl, prestwick0_000666, prestwick1_000666 | |
(2R,6S)-2,6-dimethylpiperidine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several min
GHS Signal Word: Danger
TSCA : TSCA