Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-2,6-Dimethylpiperazine, 99+%

Chemical Identifiers

Chemical Name or Material cis-2,6-Dimethylpiperazine
CAS 21655-48-1
Molecular Formula C6H14N2
Formula Weight 114.19
MDL Number MFCD07772435
UN Number UN1325
Beilstein 79881
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1568414

 
alfa aesar™
L1568414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1568406

 
alfa aesar™
L1568406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 114.19
Chemical Name or Material cis-2,6-Dimethylpiperazine
CAS 21655-48-1
Assay ≥99%
Molecular Formula C6H14N2
MDL Number MFCD07772435
UN Number UN1325
Beilstein 79881
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 25g
Flash Point 45°C (113°F)
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
Melting Point 108° to 111°C
Boiling Point 162°C
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.