Learn More
cis-2,6-Dimethylmorpholine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1300709
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
cis-2,6-Dimethylmorpholine | |
-85°C | |
146°C to 147°C | |
C6H13NO | |
MFCD00078428 | |
UN1992 | |
cis-2,6-dimethylmorpholine, 2r,6s-2,6-dimethylmorpholine, 2s,6r-2,6-dimethylmorpholine, unii-vwd860p007, 2,6-dimethylmorpholine, cis, morpholine, 2,6-dimethyl-, cis, 2,6-dimethyl-morpholine, morpholine, 2,6-dimethyl-, 2r,6s-rel, cdmm, pubchem16624 | |
CC1CNCC(O1)C | |
115.176 | |
115.18 |
6485-55-8 | |
0.935 | |
48°C (118°F) | |
1.446 | |
10 g | |
878182 | |
HNVIQLPOGUDBSU-OLQVQODUSA-N | |
(2S,6R)-2,6-dimethylmorpholine | |
641500 | |
97% |
Chemical Identifiers
6485-55-8 | |
115.176 | |
HNVIQLPOGUDBSU-OLQVQODUSA-N | |
641500 | |
CC1CNCC(O1)C |
C6H13NO | |
MFCD00078428 | |
cis-2,6-dimethylmorpholine, 2r,6s-2,6-dimethylmorpholine, 2s,6r-2,6-dimethylmorpholine, unii-vwd860p007, 2,6-dimethylmorpholine, cis, morpholine, 2,6-dimethyl-, cis, 2,6-dimethyl-morpholine, morpholine, 2,6-dimethyl-, 2r,6s-rel, cdmm, pubchem16624 | |
(2S,6R)-2,6-dimethylmorpholine |
Safety and Handling
GHS H Statement
H318-H226-H312
Causes serious eye damage.
Flammable liquid and vapor.
Harmful in contact with skin.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H226-H302-H311-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 229-353-0
RTECSNumber : QE1750000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only