Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-(1S,2R)-(-)-2-Benzylaminocyclohexanemethanol, 98%

$37.05 - $37.05

Chemical Identifiers

Chemical Name or Material cis-(1S,2R)-(-)-2-Benzylaminocyclohexanemethanol
CAS 71581-93-6
Molecular Formula C14H21NO
Formula Weight 219.33
MDL Number MFCD00151396
Beilstein 8393598
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL11749MD

  SDS
alfa aesar™
L11749MD
250mg Each for $37.05
Description & Specifications

Specifications

Formula Weight 219.33
MDL Number MFCD00151396
Optical Rotation −24° (c=1 in Methanol)
Chemical Name or Material cis-(1S,2R)-(-)-2-Benzylaminocyclohexanemethanol
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Assay 98%
Molecular Formula C14H21NO
CAS 71581-93-6
Beilstein 8393598
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 66° to 69°C
Quantity 250mg
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.