missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-(1S,2R)-(-)-2-Benzylaminocyclohexanemethanol, 98%

Manufacturer:  Alfa Aesar™ L11749MD

 View more versions of this product

Catalog No. AAL11749MD



Specifications

Specifications

cis-(1S,2R)-(-)-2-Benzylaminocyclohexanemethanol
66°C to 69°C
71581-93-6
MFCD00151396
1r,2s-n-benzyl-2-hydroxymethyl cyclohexan-1-aminium
C1CCC(C(C1)CO)NCC2=CC=CC=C2
219.328
219.33
−24° (c=1 in Methanol)
250mg
C14H21NO
8393598
BRQFIORUNWWNBM-ZIAGYGMSSA-N
[(1S,2R)-2-(benzylamino)cyclohexyl]methanol
853690
98%

Chemical Identifiers

71581-93-6
219.328
BRQFIORUNWWNBM-ZIAGYGMSSA-N
853690
C1CCC(C(C1)CO)NCC2=CC=CC=C2
C14H21NO
MFCD00151396
1r,2s-n-benzyl-2-hydroxymethyl cyclohexan-1-aminium
[(1S,2R)-2-(benzylamino)cyclohexyl]methanol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Documents