Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-(1S,2R)-(+)-2-Benzamidocyclohexanecarboxylic acid, 98%

$50.65 - $129.15

Chemical Identifiers

Chemical Name or Material cis-(1S,2R)-(+)-2-Benzamidocyclohexanecarboxylic acid
CAS 26685-82-5
Molecular Formula C14H17NO3
Formula Weight 247.3
MDL Number MFCD00078300
Beilstein 4946668
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1059403

 
alfa aesar™
L1059403
1g Each for $129.15

AAL10594MD

 
alfa aesar™
L10594MD
250mg Each for $50.65
Description & Specifications

Specifications

Formula Weight 247.3
Chemical Name or Material cis-(1S,2R)-(+)-2-Benzamidocyclohexanecarboxylic acid
Assay 98%
CAS 26685-82-5
Molecular Formula C14H17NO3
MDL Number MFCD00078300
Beilstein 4946668
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 205° to 206°C
Quantity 1g
Optical Rotation +36.5° (c=0.5 in Ethanol)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.