missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-(1S,2R)-(+)-2-Benzamidocyclohexanecarboxylic acid, 98%

Manufacturer:  Alfa Aesar™ L1059403

 View more versions of this product

Catalog No. AAL1059403



Specifications

Specifications

cis-(1S,2R)-(+)-2-Benzamidocyclohexanecarboxylic acid
205°C to 206°C
26685-82-5
MFCD00078300
1s,2r-2-benzamidocyclohexanecarboxylic acid, 1s,2r-+-2-benzamidocyclohexanecarboxylic acid, +-cis-2-benzamidocyclohexanecarboxylic acid, 1s,2r-2-benzamidocyclohexane-1-carboxylic acid, cyclohexanecarboxylic acid, 2-benzoylamino-, 1s-cis, cis-2-benzamidocyclohexane carboxylic acid, pubchem8076, +-cis-2-benzamidocyclohexanecarboxylicacid, 1s,2r-2-benzamidocyclohexanecarboxylicacid, +-1s,2r-2-benzamidocyclohexanecarboxylic acid
C1CCC(C(C1)C(=O)O)NC(=O)C2=CC=CC=C2
247.294
247.3
+36.5° (c=0.5 in Ethanol)
1g
C14H17NO3
4946668
PUANNVQABXUYKU-NWDGAFQWSA-N
(1S,2R)-2-benzamidocyclohexane-1-carboxylic acid
1268229
98%

Chemical Identifiers

26685-82-5
247.294
PUANNVQABXUYKU-NWDGAFQWSA-N
1268229
C1CCC(C(C1)C(=O)O)NC(=O)C2=CC=CC=C2
C14H17NO3
MFCD00078300
1s,2r-2-benzamidocyclohexanecarboxylic acid, 1s,2r-+-2-benzamidocyclohexanecarboxylic acid, +-cis-2-benzamidocyclohexanecarboxylic acid, 1s,2r-2-benzamidocyclohexane-1-carboxylic acid, cyclohexanecarboxylic acid, 2-benzoylamino-, 1s-cis, cis-2-benzamidocyclohexane carboxylic acid, pubchem8076, +-cis-2-benzamidocyclohexanecarboxylicacid, 1s,2r-2-benzamidocyclohexanecarboxylicacid, +-1s,2r-2-benzamidocyclohexanecarboxylic acid
(1S,2R)-2-benzamidocyclohexane-1-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents