missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-(1R,2S)-(+)-2-Benzylaminocyclohexanemethanol, 98%

Manufacturer:  Alfa Aesar™ L09603MD

 View more versions of this product

Catalog No. AAL09603MD



Specifications

Specifications

cis-(1R,2S)-(+)-2-Benzylaminocyclohexanemethanol
66°C to 69°C
71581-92-5
MFCD00151395
1r,2s-2-benzylamino cyclohexyl methanol, +-cis-2-benzylaminocyclohexanemethanol, cis-1r,2s-+-2-benzylamino cyclohexanemethanol, 1r,2s-2-benzylamino cyclohexyl methanol, --cis-2-benzylaminocyclohexanemethanol, cis-1r,2s-+-2-benzylaminocyclohexanemethanol, pubchem7939, cis-2-benzylamino cyclohexylmethanol, 2-benzylamino cyclohexyl methanol #, cis-2-benzylamino cyclohexyl methanol
C1CCC(C(C1)CO)NCC2=CC=CC=C2
219.328
219.33
+24° (c=1 in Methanol)
250mg
C14H21NO
3942893
BRQFIORUNWWNBM-KBPBESRZSA-N
[(1R,2S)-2-(benzylamino)cyclohexyl]methanol
853687
98%

Chemical Identifiers

71581-92-5
219.328
BRQFIORUNWWNBM-KBPBESRZSA-N
853687
C1CCC(C(C1)CO)NCC2=CC=CC=C2
C14H21NO
MFCD00151395
1r,2s-2-benzylamino cyclohexyl methanol, +-cis-2-benzylaminocyclohexanemethanol, cis-1r,2s-+-2-benzylamino cyclohexanemethanol, 1r,2s-2-benzylamino cyclohexyl methanol, --cis-2-benzylaminocyclohexanemethanol, cis-1r,2s-+-2-benzylaminocyclohexanemethanol, pubchem7939, cis-2-benzylamino cyclohexylmethanol, 2-benzylamino cyclohexyl methanol #, cis-2-benzylamino cyclohexyl methanol
[(1R,2S)-2-(benzylamino)cyclohexyl]methanol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Documents