missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
cis-11-Hexadecenal 95.0+%, TCI America™
$120.39 - $551.94
Chemical Identifiers
CAS | 53939-28-9 |
---|---|
Molecular Formula | C16H30O |
Molecular Weight (g/mol) | 238.415 |
MDL Number | MFCD00010124 |
InChI Key | AMTITFMUKRZZEE-WAYWQWQTSA-N |
Synonym | cis-11-Hexadecenyl Aldehyde |
PubChem CID | 5364495 |
IUPAC Name | (Z)-hexadec-11-enal |
SMILES | CCCCC=CCCCCCCCCCC=O |
Chemical Identifiers
53939-28-9 | |
238.415 | |
AMTITFMUKRZZEE-WAYWQWQTSA-N | |
5364495 | |
CCCCC=CCCCCCCCCCC=O |
C16H30O | |
MFCD00010124 | |
cis-11-Hexadecenyl Aldehyde | |
(Z)-hexadec-11-enal |
Specifications
132°C | |
1g | |
C16H30O | |
cis-11-Hexadecenyl Aldehyde | |
CCCCC=CCCCCCCCCCC=O | |
238.415 | |
238.42 | |
≥95.0% (GC) |
Yellow | |
53939-28-9 | |
MFCD00010124 | |
AMTITFMUKRZZEE-WAYWQWQTSA-N | |
(Z)-hexadec-11-enal | |
5364495 | |
Clear Liquid at 20°C | |
cis-11-Hexadecenal |
Safety and Handling
EINECSNumber : (2)-3385
RTECSNumber : MM0350000
TSCA : Yes
Recommended Storage : Refrigerator