Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-1,8-p-Menthanediol monohydrate, 98%

$37.57 - $135.15

Chemical Identifiers

Chemical Name or Material cis-1,8-p-Menthanediol monohydrate
CAS 2451-01-6
Molecular Formula C10H20O2·H2O
Formula Weight 190.28 (172.27 Anhydrous)
MDL Number MFCD00167312
Synonym Terpin monohydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1701114

  SDS
alfa aesar™
A1701114
25g Each for $37.57

AAA1701122

  SDS
alfa aesar™
A1701122
100g Each for $135.15
Description & Specifications

Specifications

Formula Weight 190.28 (172.27 Anhydrous)
Chemical Name or Material cis-1,8-p-Menthanediol monohydrate
TSCA TSCA
Assay 98%
Molecular Formula C10H20O2·H2O
MDL Number MFCD00167312
CAS 2451-01-6
Synonym Terpin monohydrate
Melting Point 114° to 117°C
Quantity 25g