missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
cis-1,4-Diacetoxy-2-butene, 96%, Thermo Scientific Chemicals
$74.40 - $233.55
Chemical Identifiers
CAS | 25260-60-0 |
---|---|
Molecular Formula | C8H12O4 |
Molecular Weight (g/mol) | 172.18 |
MDL Number | MFCD00059339,MFCD00077968 |
InChI Key | VZUAUHWZIKOMFC-ARJAWSKDSA-N |
Synonym | cis-1,4-diacetoxy-2-butene, z-but-2-ene-1,4-diyl diacetate, 2z-but-2-ene-1,4-diyl diacetate, cis-2-butene-1,4-diol diacetate, 2z-4-acetyloxybut-2-enyl acetate, 2z-4-acetyloxy but-2-en-1-yl acetate, cis-1,4-diacetoxy-2-buten, z-1,4-diacetoxy-2-butene, cis-2-buten-1,4-diol diacetate, z-but-2-ene-1,4-diyldiacetate |
PubChem CID | 643799 |
IUPAC Name | [(Z)-4-acetyloxybut-2-enyl] acetate |
SMILES | CC(=O)OC\C=C/COC(C)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1302409
|
Thermo Scientific Chemicals
L1302409 |
10 g |
Each for $74.40
|
|
|||||
AAL1302418
|
Thermo Scientific Chemicals
L1302418 |
50 g |
Each for $233.55
|
|
|||||
Description
cis-1,4-Diacetoxy-2-butene is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationscis-1,4-Diacetoxy-2-butene is used as an organic chemical synthesis intermediate.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
25260-60-0 | |
172.18 | |
VZUAUHWZIKOMFC-ARJAWSKDSA-N | |
643799 | |
CC(=O)OC\C=C/COC(C)=O |
C8H12O4 | |
MFCD00059339,MFCD00077968 | |
cis-1,4-diacetoxy-2-butene, z-but-2-ene-1,4-diyl diacetate, 2z-but-2-ene-1,4-diyl diacetate, cis-2-butene-1,4-diol diacetate, 2z-4-acetyloxybut-2-enyl acetate, 2z-4-acetyloxy but-2-en-1-yl acetate, cis-1,4-diacetoxy-2-buten, z-1,4-diacetoxy-2-butene, cis-2-buten-1,4-diol diacetate, z-but-2-ene-1,4-diyldiacetate | |
[(Z)-4-acetyloxybut-2-enyl] acetate |
Specifications
25260-60-0 | |
120°C to 121°C (18 mmHg) | |
C8H12O4 | |
MFCD00059339,MFCD00077968 | |
1724379 | |
Not miscible in water. | |
CC(=O)OC\C=C/COC(C)=O | |
172.18 | |
172.18 | |
cis-1,4-Diacetoxy-2-butene |
1.09 | |
>110°C (230°F) | |
1.4416 | |
10 g | |
cis-1,4-diacetoxy-2-butene, z-but-2-ene-1,4-diyl diacetate, 2z-but-2-ene-1,4-diyl diacetate, cis-2-butene-1,4-diol diacetate, 2z-4-acetyloxybut-2-enyl acetate, 2z-4-acetyloxy but-2-en-1-yl acetate, cis-1,4-diacetoxy-2-buten, z-1,4-diacetoxy-2-butene, cis-2-buten-1,4-diol diacetate, z-but-2-ene-1,4-diyldiacetate | |
VZUAUHWZIKOMFC-ARJAWSKDSA-N | |
[(Z)-4-acetyloxybut-2-enyl] acetate | |
643799 | |
96% |
Safety and Handling
P264b-P280-P302+P352-P332+P313-P362
H315
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only