missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-1,4-Diacetoxy-2-butene, 96%

Manufacturer:  Alfa Aesar™ L1302418

 View more versions of this product

Catalog No. AAL1302418



Description

Description

Applications
cis-1,4-Diacetoxy-2-butene is used as an organic chemical synthesis intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

cis-1,4-Diacetoxy-2-butene
120°C to 121°C (18mmHg)
25260-60-0
MFCD00059339
cis-1,4-diacetoxy-2-butene, z-but-2-ene-1,4-diyl diacetate, 2z-but-2-ene-1,4-diyl diacetate, cis-2-butene-1,4-diol diacetate, 2z-4-acetyloxybut-2-enyl acetate, 2z-4-acetyloxy but-2-en-1-yl acetate, cis-1,4-diacetoxy-2-buten, z-1,4-diacetoxy-2-butene, cis-2-buten-1,4-diol diacetate, z-but-2-ene-1,4-diyldiacetate
CC(=O)OCC=CCOC(=O)C
172.18
172.18
>110°C (230°F)
50g
C8H12O4
1724379
VZUAUHWZIKOMFC-ARJAWSKDSA-N
[(Z)-4-acetyloxybut-2-enyl] acetate
643799
96%

Chemical Identifiers

25260-60-0
172.18
VZUAUHWZIKOMFC-ARJAWSKDSA-N
643799
CC(=O)OCC=CCOC(=O)C
C8H12O4
MFCD00059339
cis-1,4-diacetoxy-2-butene, z-but-2-ene-1,4-diyl diacetate, 2z-but-2-ene-1,4-diyl diacetate, cis-2-butene-1,4-diol diacetate, 2z-4-acetyloxybut-2-enyl acetate, 2z-4-acetyloxy but-2-en-1-yl acetate, cis-1,4-diacetoxy-2-buten, z-1,4-diacetoxy-2-butene, cis-2-buten-1,4-diol diacetate, z-but-2-ene-1,4-diyldiacetate
[(Z)-4-acetyloxybut-2-enyl] acetate
Documents