Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-1,2-Dimethylcyclohexane, 99+%

Chemical Identifiers

Chemical Name or Material cis-1,2-Dimethylcyclohexane
CAS 2207-01-4
Formula Weight 112.22
MDL Number MFCD00063621
UN Number UN2263
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3146709

  SDS
alfa aesar™
3146709
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3146718

  SDS
alfa aesar™
3146718
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 112.22
Chemical Name or Material cis-1,2-Dimethylcyclohexane
Density 0.796g/mL
TSCA TSCA
Physical Form Liquid
Assay ≥99%
MDL Number MFCD00063621
CAS 2207-01-4
UN Number UN2263
Refractive Index 1.4360
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapor.
Boiling Point 129° to 130°C
Quantity 10g
Flash Point 12° (54°F)
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIMETHYLCYCLOHEXANES
Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225
Highly flammable liquid and vapor.