Learn More
cis-1,2-Dichloroethylene, 97%, stabilized, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 113380100
Specifications
cis-1, 2-Dichloroethylene | |
6°C | |
Glass bottle | |
1.28 | |
60.0°C | |
-80.0°C | |
97% | |
96% min. (GC) | |
ClCH=CHCl | |
01,188 | |
15,87 | |
KFUSEUYYWQURPO-UPHRSURJSA-N | |
(Z)-1,2-dichloroethene | |
643833 | |
96.94 | |
97% |
1.2800g/mL | |
Authentic | |
1.4475 to 1.4495 | |
0.003% MEHQ | |
Colorless | |
10mL | |
156-59-2 | |
C2H2Cl2 | |
MFCD00000925 | |
13,293 | |
acetalyne dichloride, acetylene dichloride, cis, cis-1,2-dichloroethene, cis-1,2-dichloroethylene, cis-acetylene dichloride, cis-dichloroethylene, ethene, 1,2-dichloro-, z, unii-fyo9g15jyd, z-1,2-dichloroethene, z-1,2-dichloroethylene | |
C(=CCl)Cl | |
96.94 | |
CHEBI:28805 | |
Liquid |
Chemical Identifiers
156-59-2 | |
96.94 | |
KFUSEUYYWQURPO-UPHRSURJSA-N | |
643833 | |
(Z)-1,2-dichloroethene |
C2H2Cl2 | |
MFCD00000925 | |
acetalyne dichloride, acetylene dichloride, cis, cis-1,2-dichloroethene, cis-1,2-dichloroethylene, cis-acetylene dichloride, cis-dichloroethylene, ethene, 1,2-dichloro-, z, unii-fyo9g15jyd, z-1,2-dichloroethene, z-1,2-dichloroethylene | |
CHEBI:28805 | |
C(=CCl)Cl |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
GHS P Statement:
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 205-859-7
RTECSNumber : KV9420000
TSCA : TSCA