missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-1,2-Dichloroethylene, 97%, stabilized, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 113380025

 View more versions of this product

Catalog No. AC113380025



Description

Description

Specifications

Specifications

cis-1, 2-Dichloroethylene
6°C
Glass bottle
1.28
60.0°C
-80.0°C
97%
96% min. (GC)
ClCH=CHCl
01,188
15,87
KFUSEUYYWQURPO-UPHRSURJSA-N
(Z)-1,2-dichloroethene
643833
96.94
97%
1.2800g/mL
Authentic
1.4475 to 1.4495
0.003% MEHQ
Colorless
2.5mL
156-59-2
C2H2Cl2
MFCD00000925
13,293
acetalyne dichloride, acetylene dichloride, cis, cis-1,2-dichloroethene, cis-1,2-dichloroethylene, cis-acetylene dichloride, cis-dichloroethylene, ethene, 1,2-dichloro-, z, unii-fyo9g15jyd, z-1,2-dichloroethene, z-1,2-dichloroethylene
C(=CCl)Cl
96.94
CHEBI:28805
Liquid

Chemical Identifiers

156-59-2
96.94
KFUSEUYYWQURPO-UPHRSURJSA-N
643833
(Z)-1,2-dichloroethene
C2H2Cl2
MFCD00000925
acetalyne dichloride, acetylene dichloride, cis, cis-1,2-dichloroethene, cis-1,2-dichloroethylene, cis-acetylene dichloride, cis-dichloroethylene, ethene, 1,2-dichloro-, z, unii-fyo9g15jyd, z-1,2-dichloroethene, z-1,2-dichloroethylene
CHEBI:28805
C(=CCl)Cl
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 205-859-7

RTECSNumber : KV9420000

TSCA : TSCA

SDS Documents