Please login to your online account to display your discounted pricing

cis-1,2-Dichloroethene (For EPA 500, 8000, CLP Methods)


Quantity 1x1mL
Packaging Ampule

Manufacturer: ultra scientific  HC3701

 View more versions of this product

Catalog No. US-HC-370-1

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1x1mL
Methods For EPA 500, 8000, CLP Methods
Alpha Vector Dichloroethene
Packaging Ampule
CAS 156-59-4
Chemical Name or Material cis-1, 2-Dichloroethene

Specifications

Chemical Name or Material cis-1, 2-Dichloroethene
CAS 156-59-4