missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-1,2-Dichloroethene (For EPA 500, 8000, CLP Methods)

$46.26 - $46.26

Chemical Identifiers

CAS 156-59-4
Chemical Name or Material cis-1, 2-Dichloroethene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
US-HC-370-1 Ultra Scientific
HC3701
Ampule 1x1mL Each for $46.26
Specifications Description & Specifications

Chemical Identifiers

156-59-4
cis-1, 2-Dichloroethene

Specifications

1x1mL
Dichloroethene
156-59-4
For EPA 500, 8000, CLP Methods
Ampule
cis-1, 2-Dichloroethene