missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-1,2-Cyclopentanediol, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 310290050

 View more versions of this product

Catalog No. AC310290050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

cis-1,2-Cyclopentanediol
83.0°C to 83.5°C (5.0mmHg)
Authentic
Glass bottle
C5H8(OH)2
MFCD00134192
cis-1,2-cyclopentanediol, cis-cyclopentane-1,2-diol, cis-1,2-dihydroxycyclopentane, 1r,2s-cyclopentane-1,2-diol, 1s,2r cyclopentane-1,2-diol, cis-1,2-cyclopentandiol, 1,2-cis-cyclopentanediol, cis-1,2-cyclopen-tanediol, cis-cyclopenten-1,2-diol, 1alpha,2alpha-cyclopentanediol
VCVOSERVUCJNPR-SYDPRGILSA-N
(1R,2S)-cyclopentane-1,2-diol
2733293
98%
5057-98-7
>110°C
97.5% min. (GC)
C5H10O2
1.4770 to 1.4790
5g
Solubility in water: partially soluble in water.
C1CC(C(C1)O)O
102.13
102.13

Chemical Identifiers

5057-98-7
102.13
VCVOSERVUCJNPR-SYDPRGILSA-N
2733293
C1CC(C(C1)O)O
C5H10O2
MFCD00134192
cis-1,2-cyclopentanediol, cis-cyclopentane-1,2-diol, cis-1,2-dihydroxycyclopentane, 1r,2s-cyclopentane-1,2-diol, 1s,2r cyclopentane-1,2-diol, cis-1,2-cyclopentandiol, 1,2-cis-cyclopentanediol, cis-1,2-cyclopen-tanediol, cis-cyclopenten-1,2-diol, 1alpha,2alpha-cyclopentanediol
(1R,2S)-cyclopentane-1,2-diol
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only