Please login to your online account to display your discounted pricing

cis-1,2-Cyclopentanediol, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  310290010

 View more versions of this product

Catalog No. AC310290010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid After Melting
Formula Weight 102.13
Quantity 1g
Color Clear slightly yellow
Refractive Index 1.4770 to 1.4790 (20°C, 589nm)
Molecular Formula C5H10O2
Linear Formula C5H8(OH)
CAS 5057-98-7
Chemical Name or Material cis-1, 2-Cyclopentanediol
Assay Percent Range 98%
MDL Number MFCD00134192
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material cis-1, 2-Cyclopentanediol
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H10O2
Linear Formula C5H8(OH)
Formula Weight 102.13
MDL Number MFCD00134192