Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-1,2-Cyclohexanedicarboxylic anhydride, 98%

$30.32 - $48.62

Chemical Identifiers

Chemical Name or Material cis-1,2-Cyclohexanedicarboxylic anhydride
CAS 13149-00-3
Molecular Formula C8H10O3
Formula Weight 154.17g/mol
MDL Number MFCD00005926
Beilstein 83214
Synonym cis-Hexahydrophthalic anhydride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1605318

 
alfa aesar™
A16053-18
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1605330

 
alfa aesar™
A16053-30
250g Each for $30.32

AAA1605336

 
alfa aesar™
A16053-36
500g Each for $48.62
Description & Specifications

Specifications

Melting Point 31° to 36°C
Boiling Point 296°C
Quantity 50g
Flash Point 151°C (303°F)
Chemical Name or Material cis-1,2-Cyclohexanedicarboxylic anhydride
CAS 13149-00-3
Assay 98%
Molecular Formula C8H10O3
Formula Weight 154.17g/mol
MDL Number MFCD00005926
Beilstein 83214
Synonym cis-Hexahydrophthalic anhydride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H334-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P310-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P310-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.