missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-1,2-Cyclohexanedicarboxylic acid, 98%

Manufacturer:  Alfa Aesar™ L1046614

 View more versions of this product

Catalog No. AAL1046614



Specifications

Specifications

cis-1,2-Cyclohexanedicarboxylic acid
25g
C8H12O4
2049754
QSAWQNUELGIYBC-OLQVQODUSA-N
(1S,2R)-cyclohexane-1,2-dicarboxylic acid
245564
98%
189°C to 192°C
610-09-3
MFCD00064949
cis-1,2-cyclohexanedicarboxylic acid, cis-cyclohexane-1,2-dicarboxylic acid, cis-hexahydrophthalic acid, 1r,2s-cyclohexane-1,2-dicarboxylic acid, 1s,2r-cyclohexane-1,2-dicarboxylic acid, 1,2-cyclohexanedicarboxylic acid, 1r,2s-rel, cis-1,2-cyclohexanedicarboxylicacid, ksc497o9d, cis-+/--perhydrophthalic acid, cis-cyclohexane-1,2-dicarboxylicacid
C1CCC(C(C1)C(=O)O)C(=O)O
172.18
172.18

Chemical Identifiers

610-09-3
172.18
QSAWQNUELGIYBC-OLQVQODUSA-N
245564
C1CCC(C(C1)C(=O)O)C(=O)O
C8H12O4
MFCD00064949
cis-1,2-cyclohexanedicarboxylic acid, cis-cyclohexane-1,2-dicarboxylic acid, cis-hexahydrophthalic acid, 1r,2s-cyclohexane-1,2-dicarboxylic acid, 1s,2r-cyclohexane-1,2-dicarboxylic acid, 1,2-cyclohexanedicarboxylic acid, 1r,2s-rel, cis-1,2-cyclohexanedicarboxylicacid, ksc497o9d, cis-+/--perhydrophthalic acid, cis-cyclohexane-1,2-dicarboxylicacid
(1S,2R)-cyclohexane-1,2-dicarboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents