Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-1,2-Cyclohexanedicarboxylic acid, 98%

$190.86 - $190.86

Chemical Identifiers

Chemical Name or Material cis-1,2-Cyclohexanedicarboxylic acid
CAS 610-09-3
Molecular Formula C8H12O4
Formula Weight 172.18
MDL Number MFCD00064949
Beilstein 2049754
Synonym cis-Hexahydrophthalic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1046622

 
alfa aesar™
L1046622
100g Each for $190.86
Description & Specifications

Specifications

Formula Weight 172.18
Chemical Name or Material cis-1,2-Cyclohexanedicarboxylic acid
Molecular Formula C8H12O4
MDL Number MFCD00064949
Beilstein 2049754
Synonym cis-Hexahydrophthalic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
CAS 610-09-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 189° to 192°C
Quantity 100g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.