Please login to your online account to display your discounted pricing

cis-1,2-Bis(diphenylphosphino)ethylene, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material cis-1, 2-Bis(diphenylphosphino)ethylene
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C26H22P2
Linear Formula (C6H5)2PCH=CHP(C6H5)2
Formula Weight 396.41
MDL Number MFCD00003046
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC320890010

  SDS
thermo fisher scientific
AC320890010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 396.41
Physical Form Crystals or Crystalline Needles
Quantity 1g
Color White
Melting Point 122° to 126°C
Chemical Name or Material cis-1, 2-Bis(diphenylphosphino)ethylene
Packaging Glass bottle
CAS 983-80-2
Molecular Formula C26H22P2
Assay Percent Range 98%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
MDL Number MFCD00003046
Infrared Spectrum Authentic
Linear Formula (C6H5)2PCH=CHP(C6H5)2
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.