Please login to your online account to display your discounted pricing

cis-1,2-Bis(diphenylphosphino)ethylene, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC320890010

 View more versions of this product

Catalog No. AC320890010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 396.41
Physical Form Crystals or Crystalline Needles
Quantity 1g
Color White
Melting Point 122° to 126°C
Chemical Name or Material cis-1, 2-Bis(diphenylphosphino)ethylene
Packaging Glass bottle
Molecular Formula C26H22P2
CAS 983-80-2
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula (C6H5)2PCH=CHP(C6H5)2
MDL Number MFCD00003046
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material cis-1, 2-Bis(diphenylphosphino)ethylene
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C26H22P2
Linear Formula (C6H5)2PCH=CHP(C6H5)2
Formula Weight 396.41
MDL Number MFCD00003046
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents