Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-1,2-Bis(diphenylphosphino)ethylene, 97%

$86.34 - $281.77

Chemical Identifiers

Chemical Name or Material cis-1,2-Bis(diphenylphosphino)ethylene
CAS 983-80-2
Molecular Formula C26H22P2
Formula Weight 396.41
MDL Number MFCD00003046
Beilstein 757792
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1685903

  SDS
alfa aesar™
A1685903
1g Each for $86.34

AAA1685906

  SDS
alfa aesar™
A1685906
5g Each for $281.77
Description & Specifications

Specifications

Formula Weight 396.41
Chemical Name or Material cis-1,2-Bis(diphenylphosphino)ethylene
Assay 97%
Molecular Formula C26H22P2
MDL Number MFCD00003046
CAS 983-80-2
Beilstein 757792
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 122° to 126°C
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
Intermediate in organic syntheses.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.