missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-1,2,3,6-Tetrahydrophthalic anhydride, 98%, ACROS Organics™

$45.93 - $45.93

Chemical Identifiers

CAS 935-79-5
Assay Percent Range 98%
Molecular Formula C8H8O3
Formula Weight 152.15
MDL Number MFCD00005916
Synonym cis-4-Cyclohexene-1, 2-dicarboxylic anhydride
Chemical Name or Material cis-1,2,3,6-Tetrahydrophthalic anhydride
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC159725000 SDS Thermo Fisher Scientific
159725000
Plastic bottle 500g Each for $45.93
Specifications Description & Specifications

Chemical Identifiers

935-79-5
C8H8O3
MFCD00005916
cis-1,2,3,6-Tetrahydrophthalic anhydride
98%
152.15
cis-4-Cyclohexene-1, 2-dicarboxylic anhydride
≥97.5%

Specifications

156g/mL
98° to 102°C
935-79-5
Plastic bottle
98%
152.15
17, 462
cis-1,2,3,6-Tetrahydrophthalic anhydride
White
500g
Authentic
Flakes
C8H8O3
MFCD00005916
cis-4-Cyclohexene-1, 2-dicarboxylic anhydride
≥97.5%

Safety and Handling

GHS H Statement:
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger