Please login to your online account to display your discounted pricing

cis-1,2,3,6-Tetrahydrophthalic anhydride, 98%, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  159725000

 View more versions of this product

Catalog No. AC159725000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Flakes
Formula Weight 152.15
Quantity 500g
Color White
Density 156g/mL
Melting Point 98° to 102°C
Beilstein 17, 462
Synonym cis-4-Cyclohexene-1, 2-dicarboxylic anhydride
Chemical Name or Material cis-1,2,3,6-Tetrahydrophthalic anhydride
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C8H8O3
MDL Number MFCD00005916
CAS 935-79-5
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.
Packaging Plastic bottle
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material cis-1,2,3,6-Tetrahydrophthalic anhydride
CAS 935-79-5
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H8O3
Formula Weight 152.15
MDL Number MFCD00005916
Synonym cis-4-Cyclohexene-1, 2-dicarboxylic anhydride
Accessories

Danger

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.

SDS & Safety Documents