Please login to your online account to display your discounted pricing

CIQ, Tocris™

Potentiator of NMDA receptors containing NR2C/NR2D

$218.74 - $915.10

Chemical Identifiers

Chemical Name or Material CIQ
Assay Percent Range >98%
Molecular Formula C26H26ClNO5
Formula Weight 467.94
Synonym (3-Chlorophenyl) [3, 4-dihydro-6, 7-dimethoxy-1-[(4-methoxyphenoxy)methyl]-2(1H)-isoquinolinyl]methanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

410550

 
r & d systems
RDS410550
50mg Each for $915.10

410510

 
r & d systems
RDS410510
10mg Each for $218.74
Description & Specifications

Specifications

Formula Weight 467.94
Quantity 50mg
Chemical Name or Material CIQ
Assay Percent Range >98%
Molecular Formula C26H26ClNO5
CAS 486427-17-2
Synonym (3-Chlorophenyl) [3, 4-dihydro-6, 7-dimethoxy-1-[(4-methoxyphenoxy)methyl]-2(1H)-isoquinolinyl]methanone
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C