missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cinnarizine, Thermo Scientific Chemicals
An antihistamine
Supplier: Thermo Scientific Chemicals J6456822
Description
An antihistamine
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cinnarizine | |
117°C to 121°C | |
C26H28N2 | |
100 g | |
cinnarizine, stugeron, dimitronal, stutgin, dimitron, folcodal, midronal, mitronal, stutgeron, glanil | |
DERZBLKQOCDDDZ-JLHYYAGUSA-N | |
1-benzhydryl-4-[(E)-3-phenylprop-2-enyl]piperazine | |
1547484 | |
368.52 |
298-57-7 | |
White to Yellow | |
MFCD00056037 | |
14,2305 | |
Soluble in chloroform at 50mg/ml | |
C1CN(CCN1CC=CC2=CC=CC=C2)C(C3=CC=CC=C3)C4=CC=CC=C4 | |
368.524 | |
CHEBI:31403 | |
Powder |
Chemical Identifiers
298-57-7 | |
368.524 | |
DERZBLKQOCDDDZ-JLHYYAGUSA-N | |
1547484 | |
1-benzhydryl-4-[(E)-3-phenylprop-2-enyl]piperazine |
C26H28N2 | |
MFCD00056037 | |
cinnarizine, stugeron, dimitronal, stutgin, dimitron, folcodal, midronal, mitronal, stutgeron, glanil | |
CHEBI:31403 | |
C1CN(CCN1CC=CC2=CC=CC=C2)C(C3=CC=CC=C3)C4=CC=CC=C4 |
Safety and Handling
EINECSNumber : 206-064-8
RTECSNumber : TL3430000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only