Please login to your online account to display your discounted pricing

Alfa Aesar™ Cinnamylidenemalonic acid, 98+%

$34.35 - $515.31

Chemical Identifiers

Chemical Name or Material Cinnamylidenemalonic acid
CAS 4472-92-8
Molecular Formula C12H10O4
Formula Weight 218.21
MDL Number MFCD00014345
Beilstein 1962423
Synonym 2-Carboxy-5-phenyl-2,4-pentadienoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2181109

  SDS
alfa aesar™
B2181109
10g Each for $34.35

AAB2181130

  SDS
alfa aesar™
B2181130
250g Each for $515.31
Description & Specifications

Specifications

Formula Weight 218.21
Molecular Formula C12H10O4
MDL Number MFCD00014345
Quantity 10g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay ≥98%
Health Hazard 1 Warning
CAS 4472-92-8
Melting Point ≈208°C (dec.)
Beilstein 1962423
Synonym 2-Carboxy-5-phenyl-2,4-pentadienoic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Cinnamylidenemalonic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.