missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cinnamylideneacetophenone, 98+%

Manufacturer:  Alfa Aesar™ A1511422

 View more versions of this product

Catalog No. AAA1511422



Specifications

Specifications

Cinnamylideneacetophenone
100g
C17H14O
1870052
QONKLJMPKWQQFG-HPIZBCMHSA-N
(2E,4E)-1,5-diphenylpenta-2,4-dien-1-one
1549519
≥98%
100°C to 102°C
614-57-3
MFCD00020692
cinnamylideneacetophenone, cinnamalacetophenone, 5-phenyl-2,4-pentadienophenone, 5-phenylpenta-2,4-dienophenone, 2e,4e-1,5-diphenylpenta-2,4-dien-1-one, ?,e-1,5-diphenyl-2,4-pentadien-1-one, 2,4-pentadienophenone, 5-phenyl, 2,4-pentadien-1-one, 1,5-diphenyl-, ?,e, 1,5-diphenyl-2,4-pentadien-1-one, 2,4-pentadien-1-one, 1,5-diphenyl
C1=CC=C(C=C1)C=CC=CC(=O)C2=CC=CC=C2
234.298
234.3

Chemical Identifiers

614-57-3
234.298
QONKLJMPKWQQFG-HPIZBCMHSA-N
1549519
C1=CC=C(C=C1)C=CC=CC(=O)C2=CC=CC=C2
C17H14O
MFCD00020692
cinnamylideneacetophenone, cinnamalacetophenone, 5-phenyl-2,4-pentadienophenone, 5-phenylpenta-2,4-dienophenone, 2e,4e-1,5-diphenylpenta-2,4-dien-1-one, ?,e-1,5-diphenyl-2,4-pentadien-1-one, 2,4-pentadienophenone, 5-phenyl, 2,4-pentadien-1-one, 1,5-diphenyl-, ?,e, 1,5-diphenyl-2,4-pentadien-1-one, 2,4-pentadien-1-one, 1,5-diphenyl
(2E,4E)-1,5-diphenylpenta-2,4-dien-1-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : RZ2477000

Documents