Please login to your online account to display your discounted pricing

Alfa Aesar™ Cinnamyl propionate, 97%

$74.51 - $240.12

Chemical Identifiers

Chemical Name or Material Cinnamyl propionate
CAS 103-56-0
Molecular Formula C12H14O2
Linear Formula CH3CH2CO2CH2CH=CHC6H5
Formula Weight 190.24
MDL Number MFCD00048610
Synonym Propionic acid cinnamyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2114214

 
alfa aesar™
B2114214
25g Each for $74.51

AAB2114222

 
alfa aesar™
B2114222
100g Each for $240.12
Description & Specifications

Specifications

Formula Weight 190.24
Chemical Name or Material Cinnamyl propionate
TSCA TSCA
Assay 97%
Molecular Formula C12H14O2
Linear Formula CH3CH2CO2CH2CH=CHC6H5
MDL Number MFCD00048610
CAS 103-56-0
Synonym Propionic acid cinnamyl ester
Quantity 25g
Flash Point 100°C (212°F)
RTECS Number GE2360000