missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cinnamyl chloride, 95%, stab. with sodium carbonate, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1440830

 View more versions of this product

Catalog No. AAA1440830



Description

Description

Cinnamyl chloride is used in the enantioselective total synthesis of helioporins C and E, bioactive marine diterpenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cinnamyl chloride is used in the enantioselective total synthesis of helioporins C and E, bioactive marine diterpenes.

Solubility
Soluble in water (0.2 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature: 2 - 8°C
Specifications

Specifications

Cinnamyl chloride
2687-12-9
1.096
79°C (174°F)
1.5845
250 g
741974
Soluble in water (0.2g/L at 20°C).
CCCCCCCCSCCCCCCCC
258.51
152.62
stabilized with sodium carbonate
7°C
86°C to 87°C (2 mmHg)
C16H34S
MFCD00009566
UN2927
dioctyl sulfide, di-n-octyl sulfide, octane, 1,1'-thiobis, octyl sulfide, n-octyl sulfide, dioctyl thioether, 1-octylsulfanyl octane, 9-thiaheptadecane, di-n-octylsulfide, dioctylsulfane
LOXRGHGHQYWXJK-UHFFFAOYSA-N
1-octylsulfanyloctane
75901
95%

Chemical Identifiers

2687-12-9
Safety and Handling

Safety and Handling

GHS H Statement
H330-H314-H318-H227
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P235-P260-P264b-P270-P271-P272-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c

H227-H302-H314-H317-H330-H334-H335

DOTInformation : Hazard Class: 8; Packaging Group: I

EINECSNumber : 220-246-4

RTECSNumber : CZ0919905

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only