missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cinnamyl bromide, 97%, predominantly trans, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 110400050

 View more versions of this product

Catalog No. AC110400050



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cinnamyl bromide
4392-24-9
1.3320g/mL
87°C
96% min. (HPLC)
C9H9Br
C6H5CH=CHCH2Br
5 g
1.332
RUROFEVDCUGKHD-QPJJXVBHSA-N
[(E)-3-bromoprop-1-enyl]benzene
5357478
97%
97%
26°C to 32°C
103°C (22.0 mmHg)
Authentic
Glass bottle
1.621
MFCD00000245
05, II, 372
cinnamyl bromide, 3-bromoprop-1-en-1-yl benzene, e-3-bromoprop-1-en-1-yl benzene, 3-bromo-1-phenyl-1-propene, 3-bromo-1-phenyl propene-1, e-3-bromoprop-1-enyl benzene, 3-bromo-propenyl-benzene, e-cinnamyl bromide, trans-cinnamyl bromide
C1=CC=C(C=C1)C=CCBr
197.07
197.07
Powder

Chemical Identifiers

4392-24-9
197.07
RUROFEVDCUGKHD-QPJJXVBHSA-N
5357478
C1=CC=C(C=C1)C=CCBr
C9H9Br
MFCD00000245
cinnamyl bromide, 3-bromoprop-1-en-1-yl benzene, e-3-bromoprop-1-en-1-yl benzene, 3-bromo-1-phenyl-1-propene, 3-bromo-1-phenyl propene-1, e-3-bromoprop-1-enyl benzene, 3-bromo-propenyl-benzene, e-cinnamyl bromide, trans-cinnamyl bromide
[(E)-3-bromoprop-1-enyl]benzene
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 224-511-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only