missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cinnamoyl Chloride, TCI America™

Manufacturer:  TCI America P013325G

 View more versions of this product

Catalog No. P013325G



Specifications

Specifications

Cinnamoyl Chloride
25g
C9H7ClO
3261
WOGITNXCNOTRLK-VOTSOKGWSA-N
(E)-3-phenylprop-2-enoyl chloride
5354261
Crystal-Lump at 20°C
White-Yellow
17082-09-6
MFCD00000732
cinnamoyl chloride, cinnamic acid chloride, cinnamic chloride, trans-3-phenylacryloyl chloride, 2-propenoyl chloride, 3-phenyl, cinnamoylchloride, e-cinnamoyl chloride, trans-cinnamoyl chloride, 2e-3-phenylprop-2-enoyl chloride, 3-phenyl-2-propenoyl chloride
C1=CC=C(C=C1)C=CC(=O)Cl
166.604
166.60
≥97.0% (T)

Chemical Identifiers

17082-09-6
166.604
WOGITNXCNOTRLK-VOTSOKGWSA-N
5354261
C1=CC=C(C=C1)C=CC(=O)Cl
C9H7ClO
MFCD00000732
cinnamoyl chloride, cinnamic acid chloride, cinnamic chloride, trans-3-phenylacryloyl chloride, 2-propenoyl chloride, 3-phenyl, cinnamoylchloride, e-cinnamoyl chloride, trans-cinnamoyl chloride, 2e-3-phenylprop-2-enoyl chloride, 3-phenyl-2-propenoyl chloride
(E)-3-phenylprop-2-enoyl chloride
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1720

TSCA : No

Documents