Learn More
Cinnamoyl chloride, 98%, predominantly trans, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 110395000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cinnamoyl chloride | |
102-92-1 | |
White to Yellow | |
>110°C | |
98% | |
C9H7ClO | |
MFCD00000732 | |
15, 2301 | |
Solubility in water: decomposes. Other solubilities: soluble in hydrocarbons and esters,soluble in dioxane,very soluble in logroin | |
C1=CC=C(C=C1)C=CC(=O)Cl | |
166.61 | |
166.61 | |
Crystalline Solid |
98% | |
33.0°C to 37.0°C | |
256.0°C to 258.0°C | |
Authentic | |
Glass bottle | |
C6H5CH=CHCOCl | |
500 g | |
cinnamoyl chloride, cinnamic acid chloride, cinnamic chloride, trans-3-phenylacryloyl chloride, 2-propenoyl chloride, 3-phenyl, cinnamoylchloride, e-cinnamoyl chloride, trans-cinnamoyl chloride, 2e-3-phenylprop-2-enoyl chloride, 3-phenyl-2-propenoyl chloride | |
WOGITNXCNOTRLK-VOTSOKGWSA-N | |
(E)-3-phenylprop-2-enoyl chloride | |
5354261 | |
98% |
Chemical Identifiers
102-92-1 | |
166.61 | |
WOGITNXCNOTRLK-VOTSOKGWSA-N | |
5354261 | |
C1=CC=C(C=C1)C=CC(=O)Cl |
C9H7ClO | |
MFCD00000732 | |
cinnamoyl chloride, cinnamic acid chloride, cinnamic chloride, trans-3-phenylacryloyl chloride, 2-propenoyl chloride, 3-phenyl, cinnamoylchloride, e-cinnamoyl chloride, trans-cinnamoyl chloride, 2e-3-phenylprop-2-enoyl chloride, 3-phenyl-2-propenoyl chloride | |
(E)-3-phenylprop-2-enoyl chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if inhaled.
Reacts violently with water.
Contact with water liberates toxic gas.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 203-065-5
TSCA : TSCA
RUO – Research Use Only