Please login to your online account to display your discounted pricing

Alfa Aesar™ Cinnamonitrile, cis + trans, 97%

$88.05 - $995.92

Chemical Identifiers

Chemical Name or Material Cinnamonitrile
CAS 4360-47-8
Name Note cis + trans
Molecular Formula C9H7N
Formula Weight 129.16
MDL Number MFCD00001930
Beilstein 1209545
Synonym 3-Phenylacrylonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1530114

  SDS
alfa aesar™
A1530114
25g Each for $88.05

AAA1530122

  SDS
alfa aesar™
A1530122
100g Each for $246.28

AAA1530136

  SDS
alfa aesar™
A1530136
500g Each for $995.92
Description & Specifications

Specifications

Formula Weight 129.16
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Cinnamonitrile
TSCA TSCA
Name Note cis + trans
Assay 97%
Molecular Formula C9H7N
MDL Number MFCD00001930
CAS 4360-47-8
Beilstein 1209545
Synonym 3-Phenylacrylonitrile
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 253° to 254°C
Quantity 25g
Flash Point 126°C (258°F)
RTECS Number UD1440000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.