missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cinnamonitrile, cis + trans, 97%

Manufacturer:  Alfa Aesar™ A1530122

 View more versions of this product

Catalog No. AAA1530122



Specifications

Specifications

Cinnamonitrile
253°C to 254°C
cis + trans
C9H7N
1209545
ZWKNLRXFUTWSOY-QPJJXVBHSA-N
(E)-3-phenylprop-2-enenitrile
1550846
97%
126°C (258°F)
100g
4360-47-8
MFCD00001930
cinnamonitrile, trans-cinnamonitrile, styryl cyanide, cinnamyl nitrile, 3-phenylacrylonitrile, beta-cyanostyrene, cinnamonitrile, e, 3-phenyl-2-propenenitrile, acrylonitrile, 3-phenyl, e-3-phenylprop-2-enenitrile
C1=CC=C(C=C1)C=CC#N
129.162
129.16

Chemical Identifiers

4360-47-8
129.162
ZWKNLRXFUTWSOY-QPJJXVBHSA-N
1550846
C1=CC=C(C=C1)C=CC#N
C9H7N
MFCD00001930
cinnamonitrile, trans-cinnamonitrile, styryl cyanide, cinnamyl nitrile, 3-phenylacrylonitrile, beta-cyanostyrene, cinnamonitrile, e, 3-phenyl-2-propenenitrile, acrylonitrile, 3-phenyl, e-3-phenylprop-2-enenitrile
(E)-3-phenylprop-2-enenitrile
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UD1440000

TSCA : TSCA

Documents