Learn More
Alfa Aesar™ Cinnamonitrile, cis + trans, 97%
Manufacturer: Alfa Aesar™ A1530114
Specifications
Cinnamonitrile | |
253°C to 254°C | |
cis + trans | |
C9H7N | |
1209545 | |
ZWKNLRXFUTWSOY-QPJJXVBHSA-N | |
(E)-3-phenylprop-2-enenitrile | |
1550846 | |
97% |
126°C (258°F) | |
25g | |
4360-47-8 | |
MFCD00001930 | |
cinnamonitrile, trans-cinnamonitrile, styryl cyanide, cinnamyl nitrile, 3-phenylacrylonitrile, beta-cyanostyrene, cinnamonitrile, e, 3-phenyl-2-propenenitrile, acrylonitrile, 3-phenyl, e-3-phenylprop-2-enenitrile | |
C1=CC=C(C=C1)C=CC#N | |
129.162 | |
129.16 |
Chemical Identifiers
4360-47-8 | |
129.162 | |
ZWKNLRXFUTWSOY-QPJJXVBHSA-N | |
1550846 | |
C1=CC=C(C=C1)C=CC#N |
C9H7N | |
MFCD00001930 | |
cinnamonitrile, trans-cinnamonitrile, styryl cyanide, cinnamyl nitrile, 3-phenylacrylonitrile, beta-cyanostyrene, cinnamonitrile, e, 3-phenyl-2-propenenitrile, acrylonitrile, 3-phenyl, e-3-phenylprop-2-enenitrile | |
(E)-3-phenylprop-2-enenitrile |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
RTECSNumber : UD1440000
TSCA : TSCA