Please login to your online account to display your discounted pricing

Alfa Aesar™ Cinnamamide, predominantly trans, 97%

$47.26 - $526.43

Chemical Identifiers

Chemical Name or Material Cinnamamide
CAS 621-79-4
Name Note predominantly trans
Molecular Formula C9H9NO
Formula Weight 147.18
MDL Number MFCD00008033
Beilstein 2040577
Synonym 3-Phenylacrylamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1342906

  SDS
alfa aesar™
A1342906
5g Each for $47.26

AAA1342914

  SDS
alfa aesar™
A1342914
25g Each for $153.93

AAA1342922

  SDS
alfa aesar™
A1342922
100g Each for $526.43
Description & Specifications

Specifications

Formula Weight 147.18
Chemical Name or Material Cinnamamide
TSCA TSCA
Name Note predominantly trans
Assay 97%
Molecular Formula C9H9NO
MDL Number MFCD00008033
CAS 621-79-4
Beilstein 2040577
Synonym 3-Phenylacrylamide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Melting Point 145° to 149°C
Quantity 5g
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number GD6853000
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.