missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cinnamamide, 97%, predominantly trans, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 110360250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Cinnamamide | |
621-79-4 | |
96% min. (HPLC) | |
C9H9NO | |
MFCD00008033 | |
09, 587 | |
APEJMQOBVMLION-VOTSOKGWSA-N | |
(E)-3-phenylprop-2-enamide | |
5273472 | |
147.18 |
Predominantly trans | |
Authentic | |
Glass bottle | |
C6H5CH=CHCONH2 | |
25g | |
cinnamamide, trans-cinnamamide, 3-phenylacrylamide, cinnamic amide, 2-propenamide, 3-phenyl, 3-phenylpropenamide, 2-benzylideneacetamide, e-cinnamamide, ginnamic acid amide, trans-cinnamoylamine | |
C1=CC=C(C=C1)C=CC(=O)N | |
147.18 | |
CHEBI:76320 | |
97% |
Chemical Identifiers
621-79-4 | |
147.18 | |
APEJMQOBVMLION-VOTSOKGWSA-N | |
5273472 | |
(E)-3-phenylprop-2-enamide |
C9H9NO | |
MFCD00008033 | |
cinnamamide, trans-cinnamamide, 3-phenylacrylamide, cinnamic amide, 2-propenamide, 3-phenyl, 3-phenylpropenamide, 2-benzylideneacetamide, e-cinnamamide, ginnamic acid amide, trans-cinnamoylamine | |
CHEBI:76320 | |
C1=CC=C(C=C1)C=CC(=O)N |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
EINECSNumber : 210-707-8
RUO â Research Use Only