Please login to your online account to display your discounted pricing

Cinanserin hydrochloride, Tocris


Selective 5-HT2 antagonist

Quantity 50mg

Manufacturer: r & d systems  RDS046050

 View more versions of this product

Catalog No. 046050

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 376.94
Chemical Name or Material Cinanserin hydrochloride
Assay Percent Range >99%
Molecular Formula C20H24N2OS·HCl
CAS 54-84-2
Synonym N-[2-[[3-(Dimethylamino)propyl]thio]phenyl]-3-phenyl-2-propenamide hydrochloride
Recommended Storage Desiccate at Room Temperature

Specifications

Chemical Name or Material Cinanserin hydrochloride
Assay Percent Range >99%
Molecular Formula C20H24N2OS·HCl
Formula Weight 376.94
Synonym N-[2-[[3-(Dimethylamino)propyl]thio]phenyl]-3-phenyl-2-propenamide hydrochloride
Accessories

Desiccate at Room Temperature