missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cinanserin hydrochloride, 99+%

A 5-HT antagonist

$242.11 - $874.38

Chemical Identifiers

CAS 1166-34-3
Molecular Formula C20H25ClN2OS
Molecular Weight (g/mol) 376.943
MDL Number MFCD00055609
InChI Key LXGJPDKYMJJWRB-IERUDJENSA-N
PubChem CID 6433141
IUPAC Name (E)-N-[2-[3-(dimethylamino)propylsulfanyl]phenyl]-3-phenylprop-2-enamide;hydrochloride
SMILES CN(C)CCCSC1=CC=CC=C1NC(=O)C=CC2=CC=CC=C2.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63370MA View Documents Alfa Aesar™
J63370MA
10mg Each for $242.11
 
AAJ63370MF View Documents Alfa Aesar™
J63370MF
50mg Each for $874.38
 
Specifications

Chemical Identifiers

1166-34-3
376.943
LXGJPDKYMJJWRB-IERUDJENSA-N
(E)-N-[2-[3-(dimethylamino)propylsulfanyl]phenyl]-3-phenylprop-2-enamide;hydrochloride
C20H25ClN2OS
MFCD00055609
6433141
CN(C)CCCSC1=CC=CC=C1NC(=O)C=CC2=CC=CC=C2.Cl

Specifications

White
1166-34-3
MFCD00055609
LXGJPDKYMJJWRB-IERUDJENSA-N
(E)-N-[2-[3-(dimethylamino)propylsulfanyl]phenyl]-3-phenylprop-2-enamide;hydrochloride
6433141
Solid
Cinanserin hydrochloride
10mg
C20H25ClN2OS
Soluble to 50 mM in water
CN(C)CCCSC1=CC=CC=C1NC(=O)C=CC2=CC=CC=C2.Cl
376.943
376.94
≥99%
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UC6323000