Please login to your online account to display your discounted pricing

Cinalukast, Tocris

Potent, selective CysLT1 (LTD4) antagonist; orally active

$218.74 - $915.10

Chemical Identifiers

Chemical Name or Material Cinalukast
Assay Percent Range >98%
Molecular Formula C23H28N2O3S
Formula Weight 412.55
Synonym 4-[[3-[(1E)-2-(4-Cyclobutyl-2-thiazolyl)ethenyl]phenyl]amino]-2, 2-diethyl-4-oxobutanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

202510

 
r & d systems
RDS202510
10mg Each for $218.74

202550

 
r & d systems
RDS202550
50mg Each for $915.10
Description & Specifications

Specifications

Formula Weight 412.55
Quantity 10mg
Chemical Name or Material Cinalukast
Assay Percent Range >98%
Molecular Formula C23H28N2O3S
CAS 128312-51-6
Synonym 4-[[3-[(1E)-2-(4-Cyclobutyl-2-thiazolyl)ethenyl]phenyl]amino]-2, 2-diethyl-4-oxobutanoic acid
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C