Thermo Scientific™ Cimarec+™ Stirring Hotplates Series Promotion
Precise stirring control, exceptional safety and temperature performance for your routine protocols.
$1,468.80 - $1,468.80
Specifications
Top Plate Material Ceramic
Electrical Requirements 100-120V 50/60Hz
Plug Type US-style
Includes 1 power cord
Stirring Range 50 to 1,500rpm
View More Specs
Disclaimers:
Warranty: 
Subject to 3-year warranty. See your sales representative for details
Products ${productFamilyLength}
Catalog Number Mfr. No. Dimensions (L x W x H) Max. Temperature (Metric) Surface Area (English) Heating Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W x H) Max. Temperature (Metric) Surface Area (English) Heating Price Quantity  
SP88857100P Thermo Scientific™
SP88857100PR
13 x 8.2 x 3.8 in. (33 x 20.8 x 9.7cm) 540°C 7.25 x 7.25 in. Pack of 3 Each for $1,468.80
Specifications
Description
The new Thermo Scientific™ Cimarec+™ series digital stirring hotplates offer precise stirring control, exceptional safety and temperature performance for your routine protocols. The Cimarec+ Series are available in three sizes, providing flexibility from microscale chemistries to production operations.
Advanced technology
  • Stir-Trac features innovative engineering for slow-speed stirring, consistent speed control, and strong magnetic coupling
  • Temperature adjustable in 1 degree increments
  • Hot Top warning system protects from accidental burns with prominent display when heating surface is above 50°C (122°F)
Flexible, robust design
  • Top plate choices in ceramic or aluminum
  • Easy-to-read LED display for heating and stirring
  • Raised display design protects electronics from spills
  • Optional temperature probe and splash guard protection shield
  • Variable sizes for different sample volumes
Description & Specifications
Specifications
Ceramic
US-style
50 to 1,500rpm
100-120V 50/60Hz
1 power cord
cCSAus, CE