Thermo Scientific™ Cimarec+™ Stirring Hotplate Series, Ceramic: PROMO
Get precise stirring control, exceptional safety and temperature performance for your routine protocols.
$454.75 - $655.64
Specifications
Stirring Range 50 to 1,500rpm
Certifications/Compliance cCSAus, CE
View More Specs
Products 3
Catalog Number Mfr. No. Surface Area (English) Heating Top Plate Material Electrical Requirements Plug Type Price Quantity  
Catalog Number Mfr. No. Surface Area (English) Heating Top Plate Material Electrical Requirements Plug Type Price Quantity  
SP88850100S View Documents Thermo Scientific™
SP88850100S
10.25 x 10.25 in. Ceramic 100-120V 50/60Hz US-style Each for $655.64
SP88854100S View Documents Thermo Scientific™
SP88854100S
4.25 x 4.25 in. Ceramic 100-120V 50/60Hz US-style Each for $454.75
SP88857100S View Documents Thermo Scientific™
SP88857100SD
7.25 x 7.25 in. Ceramic 100-120V 50/60Hz US-style Each for $500.72
Description
Description
Our new Thermo Scientific™ Cimarec+™ series digital stirring hotplates offer precise stirring control, exceptional safety and temperature performance for your routine protocols. The Cimarec+ Series are available in three sizes, providing flexibility from microscale chemistries to production operations.
Advanced technology Stir-Trac features innovative engineering for slow-speed stirring, consistent speed control, and strong magnetic coupling
  • Temperature adjustable in 1 degree increments
  • Hot Top warning system protects from accidental burns with prominent display when heating surface is above 50°C (122°F)
Flexible, robust design
  • Top plate choices in ceramic or aluminum
  • Easy-to-read LED display for heating and stirring
  • Raised display design protects electronics from spills
  • Optional temperature probe and splash guard protection shield
  • Variable sizes for different sample volume
Specifications
Specifications
50 to 1,500rpm
cCSAus, CE