missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cilostazol, 98%

An inhibitor of phosphodiesterase III

$158.84 - $641.53

Chemical Identifiers

CAS 73963-72-1
Molecular Formula C20H27N5O2
Molecular Weight (g/mol) 369.469
MDL Number MFCD00866780
InChI Key RRGUKTPIGVIEKM-UHFFFAOYSA-N
Synonym cilostazol, pletal, cilostazole, pletaal, cilostazolum, cilostazolum inn-latin, opc 21, opc-21, cilostazol inn:jan, pletal tn
PubChem CID 2754
ChEBI CHEBI:31401
IUPAC Name 6-[4-(1-cyclohexyltetrazol-5-yl)butoxy]-3,4-dihydro-1H-quinolin-2-one
SMILES C1CCC(CC1)N2C(=NN=N2)CCCCOC3=CC4=C(C=C3)NC(=O)CC4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62301MA View Documents Alfa Aesar™
J62301MA
10mg Each for $158.84
 
AAJ62301MF View Documents Alfa Aesar™
J62301MF
50mg Each for $641.53
 
Specifications

Chemical Identifiers

73963-72-1
369.469
RRGUKTPIGVIEKM-UHFFFAOYSA-N
2754
6-[4-(1-cyclohexyltetrazol-5-yl)butoxy]-3,4-dihydro-1H-quinolin-2-one
C20H27N5O2
MFCD00866780
cilostazol, pletal, cilostazole, pletaal, cilostazolum, cilostazolum inn-latin, opc 21, opc-21, cilostazol inn:jan, pletal tn
CHEBI:31401
C1CCC(CC1)N2C(=NN=N2)CCCCOC3=CC4=C(C=C3)NC(=O)CC4

Specifications

White
73963-72-1
MFCD00866780
cilostazol, pletal, cilostazole, pletaal, cilostazolum, cilostazolum inn-latin, opc 21, opc-21, cilostazol inn:jan, pletal tn
C1CCC(CC1)N2C(=NN=N2)CCCCOC3=CC4=C(C=C3)NC(=O)CC4
369.469
CHEBI:31401
Solid
Cilostazol
10mg
C20H27N5O2
14,2277
RRGUKTPIGVIEKM-UHFFFAOYSA-N
6-[4-(1-cyclohexyltetrazol-5-yl)butoxy]-3,4-dihydro-1H-quinolin-2-one
2754
369.46
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : VC8277500