Please login to your online account to display your discounted pricing

Cilostamide, Tocris

PDE3 inhibitor

$195.49 - $816.90

Chemical Identifiers

Chemical Name or Material Cilostamide
Assay Percent Range >98%
Molecular Formula C20H26N2O3
Formula Weight 342.44
Synonym N-Cyclohexyl-N-methyl-4-(1, 2-dihydro-2-oxo-6-quinolyloxy)butyramide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

091510

 
r & d systems
RDS091510
10mg Each for $195.49

091550

 
r & d systems
RDS091550
50mg Each for $816.90
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 342.44
Chemical Name or Material Cilostamide
Assay Percent Range >98%
Molecular Formula C20H26N2O3
CAS 68550-75-4
Synonym N-Cyclohexyl-N-methyl-4-(1, 2-dihydro-2-oxo-6-quinolyloxy)butyramide
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature