Please login to your online account to display your discounted pricing

Cilnidipine, Tocris

Ca2+ channel blocker (dual L/N-type)

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material Cilnidipine
Assay Percent Range >99%
Molecular Formula C27H28N2O7
Formula Weight 492.52
Synonym 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid 2-methoxyethyl (2E)-3-phenyl-2-propenyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

262950

 
r & d systems
RDS262950
50mg Each for $976.15

262910

 
r & d systems
RDS262910
10mg Each for $239.63
Description & Specifications

Specifications

Formula Weight 492.52
Quantity 50mg
Chemical Name or Material Cilnidipine
Recommended Storage Store at +4°C
Molecular Formula C27H28N2O7
CAS 132203-70-4
Synonym 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid 2-methoxyethyl (2E)-3-phenyl-2-propenyl ester
Assay Percent Range >99%
Safety and Handling

Store at +4°C